2-Chloro-3-fluoro-6-nitroaniline 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-3-chloro-4-fluoronitrobenzene
PC6781-1G
PC6781-5G
APOSPC6781-1GEA
705
CZK
APOSPC6781-1G
APOSPC6781-5G
2-Chloro-3-fluoro-6-nitroaniline 97%
2-Chloro-3-fluoro-6-nitroaniline
Vzorec:
C₆H₄ClFN₂O₂ MW: 190,56 g/mol Bod tání: 127…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD09763600 CAS číslo: 948014-34-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...