Tisk…

2-(2,2,2-Trifluoroethoxy)bromobenzene 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-2-(2,2,2-trifluoroethoxy)benzene

PC6679-1G PC6679-5G
APOSPC6679-1GEA 4170 CZK
APOSPC6679-1G APOSPC6679-5G
2-(2,2,2-Trifluoroethoxy)bromobenzene 95%
2-(2,2,2-Trifluoroethoxy)bromobenzene
Vzorec: C₈H₆BrF₃O
MW: 255,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09942728
CAS číslo: 218610-57-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR