2,2,2-Trifluoroethoxybenzene 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Phenoxy-1,1,1-trifluoroethane
PC6627-5G
PC6627-1G
APOSPC6627-5GEA
12960
CZK
APOSPC6627-5G
APOSPC6627-1G
2,2,2-Trifluoroethoxybenzene 95%
2,2,2-Trifluoroethoxybenzene
Vzorec:
C₈H₇F₃O MW: 176,136 g/mol Bod varu: 55…58 °C (10 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD13193045 CAS číslo: 17351-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...