Tisk…

8-Iodoperfluoro-6-oxaoctanoic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-(2-Iodo-1,1,2,2-tetrafluoroethoxy)-2,2,3,3,4,4,5,5-octafluoropentanoic acid

PC6407-1G PC6407-5G PC6407-250MG
APOSPC6407-1GEA 11540 CZK
APOSPC6407-1G APOSPC6407-5G APOSPC6407-250MG
8-Iodoperfluoro-6-oxaoctanoic acid
8-Iodoperfluoro-6-oxaoctanoic acid
Vzorec: C₇HF₁₂IO₃
Bod varu: 97 °C (4 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09763625
CAS číslo: 948014-44-6
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR