Tisk…

3,5-Difluoro-4-methoxybenzamide

Dodavatel: Apollo Scientific

PC6394-5G PC6394-1G
APOSPC6394-5GEA 34720 CZK
APOSPC6394-5G APOSPC6394-1G
3,5-Difluoro-4-methoxybenzamide
3,5-Difluoro-4-methoxybenzamide
Vzorec: C₈H₇F₂NO₂
MW: 187,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095190
CAS číslo: 343-79-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR