Tisk…

2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC6313-1G PC6313-5G PC6313-25G
APOSPC6313-1GEA 1580 CZK
APOSPC6313-1G APOSPC6313-5G APOSPC6313-25G
2-Fluoro-3-(trifluoromethoxy)benzyl bromide 95%
2-Fluoro-3-(trifluoromethoxy)benzyl bromide
Vzorec: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD12026474
CAS číslo: 1159512-59-0
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR