Tisk…

3,4-Difluorostyrene

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,2-Difluoro-4-ethenylbenzene, 1,2-Difluoro-4-vinylbenzene

PC6276-1G PC6276-25G PC6276-5G
APOSPC6276-1GEA 1080 CZK
APOSPC6276-1G APOSPC6276-25G APOSPC6276-5G
3,4-Difluorostyrene
3,4-Difluorostyrene
Vzorec: C₈H₆F₂
MW: 140.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09038471
CAS číslo: 405-03-8
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR