Tisk…

(E)-2-(Trifluoromethyl)vinyl benzyl ether

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (E)-2-(Trifluoromethyl)ethenyl benzyl ether, ({[(1E)-3,3,3-Trifluoroprop-1-en-1-yl]oxy}methyl)benzene

PC6259-25G PC6259-5G PC6259-1G
APOSPC6259-25GEA 22780 CZK
APOSPC6259-25G APOSPC6259-5G APOSPC6259-1G
(E)-2-(Trifluoromethyl)vinyl benzyl ether
(E)-2-(Trifluoromethyl)vinyl benzyl ether
Vzorec: C₁₀H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD13193051
CAS číslo: 932395-40-9
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR