6-Fluorobenzo[b]furan-3(2H)-one 98+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Fluorobenzofuran-3(2H)-one, 6-Fluoro-1-benzofuran-3(2H)-one, 2,3-Dihydro-6-fluoro-3-oxobenzo[b]furan
PC6230-1G
PC6230-250MG
PC6230-5G
APOSPC6230-1GEA
4960
CZK
APOSPC6230-1G
APOSPC6230-250MG
APOSPC6230-5G
6-Fluorobenzo[b]furan-3(2H)-one 98+%
6-Fluorobenzo[b]furan-3(2H)-one
Vzorec:
C₈H₅FO₂ MW: 152,123 g/mol Bod tání: 79…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD09743984 CAS číslo: 351528-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...