Tisk…

4-Fluorobenzo[b]furan-3(2H)-one

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluorobenzofuran-3(2H)-one, 2,3-Dihydro-4-fluorobenzo[b]furan-3-one, 4-Fluoro-2,3-dihydro-3-oxo-1-benzofuran, 4-Fluorocoumaran-3-one

PC6226-1G PC6226-250MG PC6226-5G
APOSPC6226-1GEA 15840 CZK
APOSPC6226-1G APOSPC6226-250MG APOSPC6226-5G
4-Fluorobenzo[b]furan-3(2H)-one
4-Fluorobenzo[b]furan-3(2H)-one
Vzorec: C₈H₅FO₂
MW: 152,123 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026471
CAS číslo: 911826-36-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR