Tisk…

4-(Trifluoroacetyl)benzaldehyde 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4'-Formyl-2,2,2-trifluoroacetophenone, 4-(2,2,2-trifluoroacetyl)benzaldehyde, 1-(4-Formylphenyl)-2,2,2-trifluoroethan-1-one

PC6194-1G PC6194-250MG PC6194-5G
APOSPC6194-1GEA 7480 CZK
APOSPC6194-1G APOSPC6194-250MG APOSPC6194-5G
4-(Trifluoroacetyl)benzaldehyde 95%
4-(Trifluoroacetyl)benzaldehyde
Vzorec: C₉H₅F₃O₂
MW: 202,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547943
CAS číslo: 86988-50-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR