Tisk…

4-Methyl-3-(trifluoromethyl)phenyl isocyanate 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Isocyanato-1-methyl-2-(trifluoromethyl)benzene, 5-Isocyanato-α,α,α-trifluoro-o-xylene

PC6108-1G PC6108-5G
APOSPC6108-1GEA 2200 CZK
APOSPC6108-1G APOSPC6108-5G
4-Methyl-3-(trifluoromethyl)phenyl isocyanate 97%
4-Methyl-3-(trifluoromethyl)phenyl isocyanate

May form small amount of sediment

Vzorec: C₉H₆F₃NO
MW: 201,145 g/mol
Bod varu: 190…191 °C
Hustota: 1,288 g/cm³ (25 °C)
Bod vzplanutí: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03427212
CAS číslo: 51903-64-1
UN: 2206
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR