Tisk…

Ethyl 4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate

Dodavatel: Apollo Scientific

PC6054-1G PC6054-5G
APOSPC6054-1GEA 3540 CZK
APOSPC6054-1G APOSPC6054-5G
Ethyl 4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Ethyl 4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}benzoate
Vzorec: C₁₅H₁₂F₃NO₃
MW: 311.256 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05669605
CAS číslo: 866154-36-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR