Tisk…

4-Bromo-3-(trifluoromethyl)benzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: [4-Bromo-3-(trifluoromethyl)phenyl]methylamine, 5-(Aminomethyl)-2-bromobenzotrifluoride

PC6027-1G PC6027-250MG PC6027-5G
APOSPC6027-1GEA 6020 CZK
APOSPC6027-1G APOSPC6027-250MG APOSPC6027-5G
4-Bromo-3-(trifluoromethyl)benzylamine 98%
4-Bromo-3-(trifluoromethyl)benzylamine
Vzorec: C₈H₇BrF₃N
MW: 254,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026484
CAS číslo: 1159512-70-5
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR