Tisk…

3-(Difluoromethoxy)phenylacetylene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: α,α-Difluoro-3-ethynylanisole, 1-(Difluoromethoxy)-3-ethynylbenzene, Difluoromethyl 3-ethynylphenyl ether

PC5953-1G PC5953-5G
APOSPC5953-1GEA 3740 CZK
APOSPC5953-1G APOSPC5953-5G
3-(Difluoromethoxy)phenylacetylene 98%
3-(Difluoromethoxy)phenylacetylene
Vzorec: C₉H₆F₂O
MW: 168,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094334
CAS číslo: 519059-03-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR