2-Fluoro-4-methylphenylacetonitrile 97%
Dodavatel: Apollo Scientific
Synonyms:
2-(2-Fluoro-4-methylphenyl)acetonitrile
PC5937-1G
PC5937-5G
PC5937-25G
APOSPC5937-1GEA
1580
CZK
APOSPC5937-1G
APOSPC5937-5G
APOSPC5937-25G
2-Fluoro-4-methylphenylacetonitrile 97%
2-Fluoro-4-methylphenylacetonitrile
Vzorec:
C₉H₈FN MW: 149.165 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094328 CAS číslo: 518070-26-3 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...