3-Fluoro-5-methylphenylacetonitrile 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(3-Fluoro-5-methylphenyl)acetonitrile, 3-(Cyanomethyl)-5-fluorotoluene, (3-Fluoro-5-methylphenyl)acetonitrile, 2-(3-Fluoro-5-methylphenyl)ethanonitrile
PC5919-1G
PC5919-5G
PC5919-10G
APOSPC5919-1GEA
1660
CZK
APOSPC5919-1G
APOSPC5919-5G
APOSPC5919-10G
3-Fluoro-5-methylphenylacetonitrile 97%
3-Fluoro-5-methylphenylacetonitrile
Vzorec:
C₉H₈FN MW: 149,165 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094317 CAS číslo: 518070-21-8 UN: 3276 ADR: 6.1,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...