α-Methyl-2,3,4,5,6-pentafluorophenethyl alcohol 97%
Dodavatel: Apollo Scientific
Synonyms:
1-(2-Hydroxyprop-1-yl)-2,3,4,5,6-pentafluorobenzene, 1-(Perfluorophenyl)propan-2-ol
PC5861-1G
PC5861-5G
APOSPC5861-1GEA
4240
CZK
APOSPC5861-1G
APOSPC5861-5G
α-Methyl-2,3,4,5,6-pentafluorophenethyl alcohol 97%
α-Methyl-2,3,4,5,6-pentafluorophenethyl alcohol
Vzorec:
C₉H₇F₅O MW: 226,143 g/mol Bod varu: 67…68 °C (5 mm Hg) Bod tání: 81…82 °C Bod vzplanutí: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00086181 CAS číslo: 1988-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...