Tisk…

Perfluoro(10,10-dimethyl-1-iodo-9-oxadecane)

Dodavatel: Apollo Scientific
Varování

Synonyms: 8-(Heptafluoroisopropoxy)-1-iodoperfluorooctane

PC5802-1G PC5802-5G PC5802-250MG
APOSPC5802-1GEA 3310 CZK
APOSPC5802-1G APOSPC5802-5G APOSPC5802-250MG
Perfluoro(10,10-dimethyl-1-iodo-9-oxadecane)
Perfluoro(10,10-dimethyl-1-iodo-9-oxadecane)
Vzorec: C₁₁F₂₃IO
MW: 711,985 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09763607
CAS číslo: 25080-19-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR