Tisk…

4-Amino-3-hydroxyphenylsulphur pentafluoride

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydroxy-4-(pentafluorothio)aniline

PC5798-1G PC5798-250MG
APOSPC5798-1GEA 17300 CZK
APOSPC5798-1G APOSPC5798-250MG
4-Amino-3-hydroxyphenylsulphur pentafluoride
4-Amino-3-hydroxyphenylsulphur pentafluoride
Vzorec: C₆H₆F₅NOS
MW: 235,175 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026459
CAS číslo: 1159512-48-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR