Perfluoro(1,2,3-trichlorocyclopent-1-ene)
Dodavatel: Apollo Scientific
Synonyms:
3,4,4,5,5-Pentafluoro-1,2,3-trichlorocyclopentene
PC5793-1G
PC5793-5G
APOSPC5793-1GEA
3340
CZK
APOSPC5793-1G
APOSPC5793-5G
Perfluoro(1,2,3-trichlorocyclopent-1-ene)
Perfluoro(1,2,3-trichlorocyclopent-1-ene)
Vzorec:
C₅Cl₃F₅ MW: 261,405 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00039452 CAS číslo: 3824-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...