Tisk…

4-(Trifluoromethyl)benzothiophene-2-carboxamide

Dodavatel: Apollo Scientific
Varování

PC57790-100MG PC57790-1G PC57790-250MG
APOSPC57790-100MGEA 3310 CZK
APOSPC57790-100MG APOSPC57790-1G APOSPC57790-250MG
4-(Trifluoromethyl)benzothiophene-2-carboxamide
4-(Trifluoromethyl)benzothiophene-2-carboxamide
Vzorec: C₁₀H₆F₃NOS
MW: 245,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31567874
CAS číslo: 2377846-68-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR