Tisk…

5-(Trifluoromethyl)benzothiophene-2-carboxamide

Dodavatel: Apollo Scientific
Varování

PC57787-1G PC57787-250MG
APOSPC57787-1GEA 3740 CZK
APOSPC57787-1G APOSPC57787-250MG
5-(Trifluoromethyl)benzothiophene-2-carboxamide
5-(Trifluoromethyl)benzothiophene-2-carboxamide
Vzorec: C₁₀H₆F₃NOS
MW: 245,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31567862
CAS číslo: 1609382-60-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR