Tisk…

6-(Trifluoromethyl)benzothiophene-2-carboxamide

Dodavatel: Apollo Scientific
Varování

PC57782-1G PC57782-250MG
APOSPC57782-1GEA 3740 CZK
APOSPC57782-1G APOSPC57782-250MG
6-(Trifluoromethyl)benzothiophene-2-carboxamide
6-(Trifluoromethyl)benzothiophene-2-carboxamide
Vzorec: C₁₀H₆F₃NOS
MW: 245,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31567850
CAS číslo: 2375008-67-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR