Tisk…

[3-Bromo-7-(trifluoromethyl)benzothiophen-2-yl]methanol

Dodavatel: Apollo Scientific
Varování

PC57779-1G PC57779-250MG
APOSPC57779-1GEA 7410 CZK
APOSPC57779-1G APOSPC57779-250MG
[3-Bromo-7-(trifluoromethyl)benzothiophen-2-yl]methanol
[3-Bromo-7-(trifluoromethyl)benzothiophen-2-yl]methanol
Vzorec: C₁₀H₆BrF₃OS
Storage Temperature: Ambient
MDL Number: MFCD29432909
CAS číslo: 2090924-00-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR