5-Bromo-2,3,4-trifluorobenzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC57777-1G
PC57777-250MG
PC57777-5G
APOSPC57777-1GEA
11680
CZK
APOSPC57777-1G
APOSPC57777-250MG
APOSPC57777-5G
5-Bromo-2,3,4-trifluorobenzaldehyde 95%
5-Bromo-2,3,4-trifluorobenzaldehyde
Vzorec:
C₇H₂BrF₃O MW: 238,989 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12756767 CAS číslo: 914935-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...