8-Chloro-1-fluorophenazine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Cloro-6-fluorophenazine
PC57776-1G
PC57776-250MG
PC57776-5G
APOSPC57776-1GEA
2610
CZK
APOSPC57776-1G
APOSPC57776-250MG
APOSPC57776-5G
8-Chloro-1-fluorophenazine
8-Chloro-1-fluorophenazine
Vzorec:
C₁₂H₆ClFN₂ MW: 232,641 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32070711 CAS číslo: 2407339-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...