Tisk…

2-Fluorophenazine

Dodavatel: Apollo Scientific
Varování

PC57774-1G PC57774-250MG PC57774-5G
APOSPC57774-1GEA 3060 CZK
APOSPC57774-1G APOSPC57774-250MG APOSPC57774-5G
2-Fluorophenazine
2-Fluorophenazine
Vzorec: C₁₂H₇FN₂
MW: 198,196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32070709
CAS číslo: 397-57-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR