3-Bromo-5-(difluoromethyl)benzoic acid 95%
Dodavatel: Apollo Scientific
PC57725-1G
PC57725-250MG
APOSPC57725-1GEA
29760
CZK
APOSPC57725-1G
APOSPC57725-250MG
3-Bromo-5-(difluoromethyl)benzoic acid 95%
3-Bromo-5-(difluoromethyl)benzoic acid
Vzorec:
C₈H₅BrF₂O₂ MW: 251.025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32070683 CAS číslo: 2169047-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...