2-Bromo-4-(difluoromethyl)benzoic acid 95%
Dodavatel: Apollo Scientific
PC57721-1G
PC57721-250MG
APOSPC57721-1GEA
20470
CZK
APOSPC57721-1G
APOSPC57721-250MG
2-Bromo-4-(difluoromethyl)benzoic acid 95%
2-Bromo-4-(difluoromethyl)benzoic acid
Vzorec:
C₈H₅BrF₂O₂ MW: 251.025 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32070680 CAS číslo: 1784852-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...