Tisk…

2-Chloro-1,1,1,3-tetrafluorobut-2-ene 97%

Dodavatel: Apollo Scientific

PC5759-1G PC5759-5G
APOSPC5759-1GEA 3070 CZK
APOSPC5759-1G APOSPC5759-5G
2-Chloro-1,1,1,3-tetrafluorobut-2-ene 97%
2-Chloro-1,1,1,3-tetrafluorobut-2-ene
Vzorec: C₄H₃ClF₄
MW: 162,513 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461615
CAS číslo: 885276-06-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR