Tisk…

2-Hydroxy-5-(trifluoromethyl)benzoic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Carboxy-4-hydroxybenzotrifluoride, 5-(Trifluoromethyl)salicylic acid, 2-Carboxy-4-(trifluoromethyl)phenol

PC5736-1G PC5736-250MG PC5736-5G
APOSPC5736-1GEA 3810 CZK
APOSPC5736-1G APOSPC5736-250MG APOSPC5736-5G
2-Hydroxy-5-(trifluoromethyl)benzoic acid 98%
2-Hydroxy-5-(trifluoromethyl)benzoic acid
Vzorec: C₈H₅F₃O₃
MW: 206.119 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07787570
CAS číslo: 79427-88-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR