Tisk…

2,3-Dibromo-1H,1H,1H,3H-perfluorobutane 97%

Dodavatel: Apollo Scientific

Synonyms: 2,3-Dibromo-1,1,1,3-tetrafluorobutane

PC5727-25G PC5727-5G
APOSPC5727-25GEA 13430 CZK
APOSPC5727-25G APOSPC5727-5G
2,3-Dibromo-1H,1H,1H,3H-perfluorobutane 97%
2,3-Dibromo-1H,1H,1H,3H-perfluorobutane
Vzorec: C₄H₄Br₂F₄
MW: 287,876 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460472
CAS číslo: 933600-85-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR