1,2-Dibromo-1H,1H,3H,3H-perfluorobutane 97%
Dodavatel: Apollo Scientific
Synonyms:
3,4-Dibromo-1,1,1,3-tetrafluorobutane
PC5726-25G
PC5726-5G
APOSPC5726-25GEA
10450
CZK
APOSPC5726-25G
APOSPC5726-5G
1,2-Dibromo-1H,1H,3H,3H-perfluorobutane 97%
1,2-Dibromo-1H,1H,3H,3H-perfluorobutane
Vzorec:
C₄H₄Br₂F₄ MW: 287,876 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08460471 CAS číslo: 933668-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...