Tisk…

Ethyl 2-hydroxy-3,3,3-trifluoropropanoate 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Ethyl 2-hydroxy-3,3,3-trifluoropropionate

PC5713-25G PC5713-5G
APOSPC5713-25GEA 42650 CZK
APOSPC5713-25G APOSPC5713-5G
Ethyl 2-hydroxy-3,3,3-trifluoropropanoate 96%
Ethyl 2-hydroxy-3,3,3-trifluoropropanoate
Vzorec: C₅H₇F₃O₃
MW: 172,102 g/mol
Bod varu: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664163
CAS číslo: 94726-00-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR