Tisk…

Potassium trifluoromethanesulphinate

Dodavatel: Apollo Scientific
Varování

PC5712-5G PC5712-1G
APOSPC5712-5GEA 21660 CZK
APOSPC5712-5G APOSPC5712-1G
Potassium trifluoromethanesulphinate
Potassium trifluoromethanesulphinate
Vzorec: CF₃O₂S·K
Bod tání: 170…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00054650
CAS číslo: 41804-89-1
EINECS: 255-556-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR