4-Bromo-3-chloro-2-fluorobenzyl alcohol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC57076-1G
PC57076-250MG
PC57076-5G
APOSPC57076-1GEA
3150
CZK
APOSPC57076-1G
APOSPC57076-250MG
APOSPC57076-5G
4-Bromo-3-chloro-2-fluorobenzyl alcohol 95%
4-Bromo-3-chloro-2-fluorobenzyl alcohol
Vzorec:
C₇H₅BrClFO Storage Temperature: Ambient |
MDL Number:
MFCD28142598 CAS číslo: 1702154-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...