Tisk…

5’-Bromo-2’-hydroxy-2,2,2-trifluoroacetophenone 99%

Dodavatel: Apollo Scientific
Varování

PC57074-1G PC57074-250MG PC57074-5G
APOSPC57074-1GEA 6250 CZK
APOSPC57074-1G APOSPC57074-250MG APOSPC57074-5G
5’-Bromo-2’-hydroxy-2,2,2-trifluoroacetophenone 99%
5’-Bromo-2’-hydroxy-2,2,2-trifluoroacetophenone
Vzorec: C₈H₄BrF₃O₂
MW: 269,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27665753
CAS číslo: 1190130-82-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR