Tisk…

4-Bromo-2,6-difluoropyridine

Dodavatel: Apollo Scientific
Varování

PC57043-1G PC57043-250MG PC57043-5G
APOSPC57043-1GEA 8440 CZK
APOSPC57043-1G APOSPC57043-250MG APOSPC57043-5G
4-Bromo-2,6-difluoropyridine
4-Bromo-2,6-difluoropyridine
Vzorec: C₅H₂BrF₂N
Storage Temperature: Refrigerator
MDL Number: MFCD21603855
CAS číslo: 903513-58-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR