2-(Methylsulphonyl)-5-(trifluoromethyl)benzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Cyano-4-(methylsulphonyl)benzotrifluoride, 2-Cyano-4-(trifluoromethyl)phenyl methyl sulphone
PC57024-1G
PC57024-5G
PC57024-10G
APOSPC57024-1GEA
1740
CZK
APOSPC57024-1G
APOSPC57024-5G
APOSPC57024-10G
2-(Methylsulphonyl)-5-(trifluoromethyl)benzonitrile
2-(Methylsulphonyl)-5-(trifluoromethyl)benzonitrile
Vzorec:
C₉H₆F₃NO₂S MW: 249,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21606312 CAS číslo: 1361385-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...