Methyl 2-chloro-5-fluoroisonicotinate
Dodavatel: Apollo Scientific
|
Varování
|
PC57011-250MG
PC57011-1G
PC57011-100MG
APOSPC57011-250MGEA
2650
CZK
APOSPC57011-250MG
APOSPC57011-1G
APOSPC57011-100MG
Methyl 2-chloro-5-fluoroisonicotinate
Methyl 2-chloro-5-fluoroisonicotinate
Vzorec:
C₇H₅ClFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09909836 CAS číslo: 876919-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...