Tisk…

3-Bromo-2-fluoro-6-iodoaniline 95%

Dodavatel: Apollo Scientific
Varování

PC56999-1G PC56999-250MG
APOSPC56999-1GEA 20970 CZK
APOSPC56999-1G APOSPC56999-250MG
3-Bromo-2-fluoro-6-iodoaniline 95%
3-Bromo-2-fluoro-6-iodoaniline
Vzorec: C₆H₄BrFIN
Storage Temperature: Ambient
MDL Number: MFCD31443828
CAS číslo: 2090537-24-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR