Tisk…

2-Bromo-6-chloro-4-(trifluoromethyl)benzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC56967-1G PC56967-250MG PC56967-5G
APOSPC56967-1GEA 953 CZK
APOSPC56967-1G APOSPC56967-250MG APOSPC56967-5G
2-Bromo-6-chloro-4-(trifluoromethyl)benzoic acid 95%
2-Bromo-6-chloro-4-(trifluoromethyl)benzoic acid
Vzorec: C₈H₃BrClF₃O₂
MW: 303,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28991853
CAS číslo: 1956322-96-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR