Tisk…

2-Fluoro-4-chlorophenacyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-1-(4-chloro-2-fluorophenyl)ethanone

PC56955-1G PC56955-250MG PC56955-5G
APOSPC56955-1GEA 910 CZK
APOSPC56955-1G APOSPC56955-250MG APOSPC56955-5G
2-Fluoro-4-chlorophenacyl bromide 97%
2-Fluoro-4-chlorophenacyl bromide
Vzorec: C₈H₅BrClFO
MW: 251,48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194236
CAS číslo: 725743-41-9
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR