Tisk…

4-Bromo-2,6-difluoro-N-methylbenzenesulphonamide

Dodavatel: Apollo Scientific
Varování

PC56954-1G PC56954-250MG PC56954-5G
APOSPC56954-1GEA 8900 CZK
APOSPC56954-1G APOSPC56954-250MG APOSPC56954-5G
4-Bromo-2,6-difluoro-N-methylbenzenesulphonamide
4-Bromo-2,6-difluoro-N-methylbenzenesulphonamide
Vzorec: C₇H₆BrF₂NO₂S
MW: 286,094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18426223
CAS číslo: 1263274-99-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR