Tisk…

2-Amino-6-(difluoromethyl)pyridine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-(Difluoromethyl)pyridin-2-amine

PC56952-100MG PC56952-1G PC56952-250MG
APOSPC56952-100MGEA 1240 CZK
APOSPC56952-100MG APOSPC56952-1G APOSPC56952-250MG
2-Amino-6-(difluoromethyl)pyridine 98%
2-Amino-6-(difluoromethyl)pyridine
Vzorec: C₆H₆F₂N₂
MW: 144,122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25476529
CAS číslo: 1315611-68-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR