Tisk…

4-(1,1,1,2,3,3,3-Heptafluoropropan-2-yl)-2-methylphenol 95%

Dodavatel: Apollo Scientific
Varování

PC56944-1G PC56944-250MG PC56944-5G
APOSPC56944-1GEA 16340 CZK
APOSPC56944-1G APOSPC56944-250MG APOSPC56944-5G
4-(1,1,1,2,3,3,3-Heptafluoropropan-2-yl)-2-methylphenol 95%
4-(1,1,1,2,3,3,3-Heptafluoropropan-2-yl)-2-methylphenol
Vzorec: C₁₀H₇F₇O
MW: 276,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31700004
CAS číslo: 353273-04-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR