Tisk…

4-Chloro-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

PC56940-1G PC56940-250MG
APOSPC56940-1GEA 27910 CZK
APOSPC56940-1G APOSPC56940-250MG
4-Chloro-2-(trifluoromethyl)pyrimidine-5-carboxylic acid 95%
4-Chloro-2-(trifluoromethyl)pyrimidine-5-carboxylic acid
Vzorec: C₆H₂ClF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11558920
CAS číslo: 1076197-55-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR