Tisk…

2-[(2,4-Dimethoxyphenyl)methylamino]-5-(trifluoromethyl)benzenesulphonamide 95%

Dodavatel: Apollo Scientific
Varování

PC56939-1G PC56939-250MG
APOSPC56939-1GEA 18620 CZK
APOSPC56939-1G APOSPC56939-250MG
2-[(2,4-Dimethoxyphenyl)methylamino]-5-(trifluoromethyl)benzenesulphonamide 95%
2-[(2,4-Dimethoxyphenyl)methylamino]-5-(trifluoromethyl)benzenesulphonamide
Vzorec: C₁₆H₁₇F₃N₂O₄S
MW: 390,377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695610
CAS číslo: 2366994-37-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR