Tisk…

(1-Fluorocyclobutyl)methanamine hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

PC56934-100MG PC56934-1G PC56934-250MG
APOSPC56934-100MGEA 18620 CZK
APOSPC56934-100MG APOSPC56934-1G APOSPC56934-250MG
(1-Fluorocyclobutyl)methanamine hydrochloride 95%
(1-Fluorocyclobutyl)methanamine hydrochloride
Vzorec: C₅H₁₀FN·ClH
MW: 139,599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27922430
CAS číslo: 1462885-81-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR